Assay Details
Target Gene Details
Entrez Gene ID: | 20533 |
Gene Name: | solute carrier family 4 (anion exchanger), member 1 |
Gene Aliases: |
Ae1, CD233, Empb3, l11Jus51 |
Location: |
Chr.11:102348820-102365281 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc4a1 | NM_011403.2 | 11 | 1480 | NP_035533.1 |
| AK010967.1 | ||||
| AK137542.1 | 11 | 1376 | BAE23401.1 | |
| AK157181.1 | 11 | 1449 | BAE33990.1 | |
| AK157833.1 | 11 | 1338 | BAE34217.1 | |
| AK157861.1 | 11 | 1350 | BAE34239.1 | |
| AK158143.1 | 11 | 1481 | BAE34381.1 | |
| AK158184.1 | 11 | 1446 | BAE34398.1 | |
| AK158243.1 | 11 | 1350 | BAE34420.1 | |
| AK158453.1 | 11 | 1325 | BAE34515.1 | |
| AK162616.1 | 11 | 1325 | BAE36992.1 | |
| AK165195.1 | 11 | 1337 | BAE38072.1 | |
| AK165267.1 | 11 | 1343 | BAE38112.1 | |
| AK171808.1 | 11 | 1314 | BAE42674.1 | |
| AK172103.1 | 11 | 1337 | ||
| AK172316.1 | 11 | 1325 | BAE42941.1 | |
| AK172395.1 | 11 | 1376 | BAE42983.1 | |
| AK172456.1 | 11 | 1360 | ||
| AK172569.1 | 11 | 1325 | BAE43071.1 | |
| AY296129.1 | 11 | 1313 | AAP51174.1 | |
| BC052419.1 | 11 | 1320 | AAH52419.1 | |
| BC053429.1 | 11 | 1311 | AAH53429.1 | |
| M29379.1 | 11 | 1307 | AAA37187.1 | |
| X02677.1 | 11 | 1313 | CAA26506.1 | |
| X03917.1 | 9 | 1157 | CAA27555.1 |
More Information
Set Membership: |
|

Genomic Map