Assay Details
Target Gene Details
Entrez Gene ID: | 637515 |
Gene Name: | NLR family, pyrin domain containing 1B |
Gene Aliases: |
ENSMUSG00000070390, Nalp1b, OTTMUSG00000006087, OTTMUSG00000006089 |
Location: |
Chr.11:71153014-71230774 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nlrp1b | NM_001040696.1 | NP_001035786.1 | ||
| NM_001162414.1 | NP_001155886.1 | |||
| BC141354.1 | ||||
| DQ117584.1 | AAZ40510.1 | |||
| DQ117585.1 | AAZ40511.1 | |||
| DQ117586.1 | AAZ40512.1 | |||
| DQ117587.1 | AAZ40513.1 | |||
| DQ117588.1 | AAZ40514.1 | |||
| DQ117589.1 | AAZ40515.1 | |||
| DQ117590.1 | AAZ40516.1 | |||
| DQ117591.1 | AAZ40517.1 | |||
| DQ117592.1 | AAZ40518.1 | |||
| DQ117593.1 | AAZ40519.1 | |||
| DQ117594.1 | AAZ40520.1 | |||
| DQ117595.1 | AAZ40521.1 | |||
| DQ117596.1 | AAZ40522.1 | |||
| DQ117597.1 | AAZ40523.1 | |||
| DQ117598.1 | AAZ40524.1 | |||
| DQ117599.1 | AAZ40525.1 | |||
| DQ117600.1 | AAZ40526.1 | |||
| DQ153214.1 | AAZ40529.1 | |||
| DQ153215.1 | AAZ40530.1 | |||
| DQ860103.1 | ||||
| DQ860104.1 | ||||
| EF158039.1 |

Genomic Map