Assay Details
Target Gene Details
Entrez Gene ID: | 209586 |
Gene Name: | NudC domain containing 3 |
Gene Aliases: |
AI427847, BC024322, mKIAA1068 |
Location: |
Chr.11:6105691-6200862 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudcd3 | NM_173748.4 | 3 | 899 | NP_776109.1 |
| AK030050.1 | 3 | 625 | ||
| AK036694.1 | 3 | 604 | ||
| AK046219.1 | 3 | 621 | BAC32642.1 | |
| AK049739.1 | 3 | 611 | BAC33899.1 | |
| AK050476.1 | 3 | 589 | BAC34277.1 | |
| AK159391.1 | 3 | 631 | BAE35045.1 | |
| AK159458.1 | 3 | 631 | BAE35100.1 | |
| AK169797.1 | 3 | 635 | BAE41373.1 | |
| AK170329.1 | 3 | 632 | BAE41723.1 | |
| AK171224.1 | 3 | 614 | BAE42326.1 | |
| AK198358.1 | 2 | 131 | ||
| BC024322.1 | 3 | 617 | AAH24322.3 | |
| BC037090.1 | 3 | 598 | AAH37090.1 | |
| BC057603.1 | 2 | 367 | AAH57603.1 | |
| EU350933.1 | 3 | 590 |
More Information
Set Membership: |
|

Genomic Map