Assay Details
Target Gene Details
Entrez Gene ID: | 209586 |
Gene Name: | NudC domain containing 3 |
Gene Aliases: |
AI427847, BC024322, mKIAA1068 |
Location: |
Chr.11:6105691-6200862 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudcd3 | NM_173748.4 | NP_776109.1 | ||
| AK030050.1 | ||||
| AK036694.1 | ||||
| AK046219.1 | BAC32642.1 | |||
| AK049739.1 | BAC33899.1 | |||
| AK050476.1 | BAC34277.1 | |||
| AK159391.1 | BAE35045.1 | |||
| AK159458.1 | BAE35100.1 | |||
| AK169797.1 | BAE41373.1 | |||
| AK170329.1 | BAE41723.1 | |||
| AK171224.1 | BAE42326.1 | |||
| AK198358.1 | ||||
| BC024322.1 | AAH24322.3 | |||
| BC037090.1 | AAH37090.1 | |||
| BC057603.1 | AAH57603.1 | |||
| EU350933.1 |
More Information
Set Membership: |
|

Genomic Map