Assay Details
Target Gene Details
Entrez Gene ID: | 209586 |
Gene Name: | NudC domain containing 3 |
Gene Aliases: |
AI427847, BC024322, mKIAA1068 |
Location: |
Chr.11:6105691-6200862 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudcd3 | NM_173748.4 | 5 | 1247 | NP_776109.1 |
| AK030050.1 | 5 | 973 | ||
| AK036694.1 | 5 | 952 | ||
| AK049739.1 | 5 | 959 | BAC33899.1 | |
| AK050476.1 | 4 | 793 | BAC34277.1 | |
| AK129276.1 | 2 | 162 | BAC98086.1 | |
| AK159391.1 | 5 | 979 | BAE35045.1 | |
| AK159458.1 | 5 | 979 | BAE35100.1 | |
| AK169797.1 | 5 | 983 | BAE41373.1 | |
| AK170329.1 | 5 | 980 | BAE41723.1 | |
| AK171224.1 | 5 | 962 | BAE42326.1 | |
| AK177508.1 | 1 | 154 | ||
| AK186218.1 | 1 | 70 | ||
| AK204837.1 | 2 | 154 | ||
| AK208209.1 | 2 | 154 | ||
| BC024322.1 | 5 | 965 | AAH24322.3 | |
| BC037090.1 | 5 | 946 | AAH37090.1 | |
| BC057603.1 | 4 | 715 | AAH57603.1 | |
| EU350933.1 | 5 | 938 |
More Information
Set Membership: |
|

Genomic Map