Assay Details
Target Gene Details
Entrez Gene ID: | 217371 |
Gene Name: | Rab40B, member RAS oncogene family |
Gene Aliases: |
SEC4L, rar |
Location: |
Chr.11:121356121-121388251 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab40b | NM_139147.3 | NP_631886.2 | ||
| AB232638.1 | BAF02900.1 | |||
| AF425643.1 | AAL60520.1 | |||
| BB847804.1 | ||||
| BC096601.1 | AAH96601.1 | |||
| BY138215.1 | ||||
| BY261509.1 | ||||
| BY283181.1 |

Genomic Map