Assay Details
Target Gene Details
Entrez Gene ID: | 64934 |
Gene Name: | pescadillo homolog 1, containing BRCT domain (zebrafish) |
Gene Aliases: |
- |
Location: |
Chr.11:3963975-3980004 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pes1 | NM_022889.3 | NP_075027.1 | ||
| AF289539.1 | AAG40734.1 | |||
| AK029075.1 | BAC26278.1 | |||
| AK088831.1 | BAC40599.1 | |||
| AK184561.1 | ||||
| BC004844.1 | AAH04844.1 | |||
| BC011142.1 | AAH11142.1 |
More Information
Set Membership: |
|

Genomic Map