Assay Details
Target Gene Details
Entrez Gene ID: | 54189 |
Gene Name: | rabaptin, RAB GTPase binding effector protein 1 |
Gene Aliases: |
Rab5ep, rabaptin-5, rabaptin-5alpha |
Location: |
Chr.11:70844723-70943105 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rabep1 | NM_001291141.1 | 2 | 302 | NP_001278070.1 |
| NM_001291142.1 | 3 | 431 | NP_001278071.1 | |
| NM_001291143.1 | 4 | 502 | NP_001278072.1 | |
| NM_019400.3 | 3 | 431 | NP_062273.2 | |
| AB001750.1 | 4 | 338 | BAA23818.1 | |
| AF248489.1 | 3 | 217 | AAF78238.1 | |
| AF248490.1 | 2 | 88 | AAG44544.1 | |
| AK019413.1 | 3 | 405 | BAB31710.3 | |
| AK134587.1 | 3 | 430 | ||
| AK137236.1 | 3 | 396 | ||
| AK151901.1 | 3 | 400 | BAE30784.1 | |
| BC060166.1 | 3 | 410 | AAH60166.1 | |
| BC129853.1 | 3 | 250 | ||
| BC129854.1 | 3 | 250 | ||
| BY718394.1 | 3 | 430 | ||
| D86066.1 | 3 | 267 | BAA21783.1 |

Genomic Map