Assay Details
Target Gene Details
Entrez Gene ID: | 19069 |
Gene Name: | nucleoporin 88 |
Gene Aliases: |
Nup84, Prei2 |
Location: |
Chr.11:70943058-70969973 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nup88 | NM_001083331.2 | 17 | 2298 | NP_001076800.1 |
| NM_001276406.1 | 17 | 2280 | NP_001263335.1 | |
| NM_172394.3 | 17 | 2265 | NP_765982.2 | |
| NR_075087.1 | 15 | 2605 | ||
| AJ532593.1 | CAD58713.1 | |||
| AK002643.1 | 17 | 2266 | ||
| AK028563.1 | 17 | 2288 | BAC26010.1 | |
| AK050893.1 | 13 | 2816 | BAC34449.1 | |
| AK139738.1 | 15 | 2513 | ||
| AK160619.1 | 17 | 2266 | BAE35919.1 | |
| AK166214.1 | 17 | 2270 | BAE38633.1 | |
| BB442284.2 | 2 | 307 | ||
| BC032929.1 | 17 | 2246 | AAH32929.1 | |
| U01135.1 | 5 | 487 |
More Information
Set Membership: |
|

Genomic Map