Assay Details
Target Gene Details
Entrez Gene ID: | 19192 |
Gene Name: | proteaseome (prosome, macropain) activator subunit 3 (PA28 gamma, Ki) |
Gene Aliases: |
AA410043, AU020960, Ki, PA28gamma, REGgamma, pa28g |
Location: |
Chr.11:101316251-101323531 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psme3 | NM_011192.3 | NP_035322.1 | ||
| AK140597.1 | BAE24425.1 | |||
| AK150939.1 | BAE29974.1 | |||
| AK156079.1 | BAE33577.1 | |||
| AK171348.1 | BAE42405.1 | |||
| BC005524.1 | AAH05524.1 | |||
| BC015255.1 | AAH15255.1 | |||
| BC139818.1 | ||||
| CT010198.1 | CAJ18406.1 | |||
| D87911.1 | BAA22041.1 | |||
| U60330.1 | AAC53297.1 |

Genomic Map