Assay Details
Target Gene Details
Entrez Gene ID: | 54189 |
Gene Name: | rabaptin, RAB GTPase binding effector protein 1 |
Gene Aliases: |
Rab5ep, rabaptin-5, rabaptin-5alpha |
Location: |
Chr.11:70844723-70943105 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rabep1 | NM_001291141.1 | 6 | 948 | NP_001278070.1 |
| NM_001291142.1 | 6 | 957 | NP_001278071.1 | |
| NM_001291143.1 | 8 | 1148 | NP_001278072.1 | |
| NM_019400.3 | 7 | 1077 | NP_062273.2 | |
| AB001750.1 | 8 | 984 | BAA23818.1 | |
| AF248489.1 | 6 | 743 | AAF78238.1 | |
| AF248490.1 | 6 | 734 | AAG44544.1 | |
| AK019413.1 | 7 | 1051 | BAB31710.3 | |
| AK134587.1 | 7 | 1077 | ||
| AK137236.1 | 7 | 1042 | ||
| BC060166.1 | 7 | 1056 | AAH60166.1 | |
| BC129853.1 | 7 | 896 | ||
| BC129854.1 | 7 | 896 | ||
| D86066.1 | 7 | 913 | BAA21783.1 |

Genomic Map