Assay Details
Target Gene Details
Entrez Gene ID: | 27681 |
Gene Name: | SNF8, ESCRT-II complex subunit, homolog (S. cerevisiae) |
Gene Aliases: |
D11Moh34 |
Location: |
Chr.11:96034890-96047430 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snf8 | NM_033568.2 | NP_291046.1 | ||
| AK013058.1 | BAB28626.1 | |||
| BC003938.1 | AAH03938.1 |

Genomic Map