Assay Details
Target Gene Details
Entrez Gene ID: | 19069 |
Gene Name: | nucleoporin 88 |
Gene Aliases: |
Nup84, Prei2 |
Location: |
Chr.11:70943058-70969973 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nup88 | NM_001083331.2 | NP_001076800.1 | ||
| NM_001276406.1 | NP_001263335.1 | |||
| NM_172394.3 | NP_765982.2 | |||
| NR_075087.1 | ||||
| AJ532593.1 | CAD58713.1 | |||
| AK002643.1 | ||||
| AK028563.1 | BAC26010.1 | |||
| AK050893.1 | BAC34449.1 | |||
| AK051122.1 | ||||
| AK085776.1 | ||||
| AK139738.1 | ||||
| AK160619.1 | BAE35919.1 | |||
| AK166214.1 | BAE38633.1 | |||
| BC032929.1 | AAH32929.1 |
More Information
Set Membership: |
|

Genomic Map