Assay Details
Target Gene Details
Entrez Gene ID: | 19069 |
Gene Name: | nucleoporin 88 |
Gene Aliases: |
Nup84, Prei2 |
Location: |
Chr.11:70943058-70969973 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nup88 | NM_001083331.2 | 2 | 410 | NP_001076800.1 |
| NM_001276406.1 | 2 | 410 | NP_001263335.1 | |
| NM_172394.3 | 2 | 410 | NP_765982.2 | |
| NR_075087.1 | 2 | 972 | ||
| AJ532593.1 | 2 | 404 | CAD58713.1 | |
| AK002643.1 | 2 | 396 | ||
| AK028563.1 | 2 | 400 | BAC26010.1 | |
| AK039007.1 | 2 | 400 | BAC30201.1 | |
| AK050893.1 | 2 | 399 | BAC34449.1 | |
| AK085776.1 | 2 | 411 | ||
| AK139738.1 | 2 | 880 | ||
| AK151739.1 | 2 | 400 | BAE30652.1 | |
| AK160619.1 | 2 | 411 | BAE35919.1 | |
| AK166214.1 | 2 | 400 | BAE38633.1 | |
| BC032929.1 | 2 | 391 | AAH32929.1 |
More Information
Set Membership: |
|

Genomic Map