Assay Details
Target Gene Details
Entrez Gene ID: | 56012 |
Gene Name: | phosphoglycerate mutase 2 |
Gene Aliases: |
- |
Location: |
Chr.11:5801637-5803796 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pgam2 | NM_018870.3 | 3 | 856 | NP_061358.1 |
| AF029843.1 | 3 | 793 | AAC13263.1 | |
| AK146703.1 | 3 | 783 | BAE27370.1 | |
| AK169019.1 | 3 | 783 | BAE40816.1 | |
| AV041299.2 | 2 | 240 | ||
| BC010750.1 | 3 | 761 | AAH10750.1 |
More Information
Set Membership: |
|

Genomic Map