Assay Details
Target Gene Details
Entrez Gene ID: | 19360 |
Gene Name: | RAD50 homolog (S. cerevisiae) |
Gene Aliases: |
Mrell, Rad50l |
Location: |
Chr.11:53649519-53707319 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rad50 | NM_009012.2 | NP_033038.2 | ||
| AK018001.1 | BAB31030.1 | |||
| AK037641.1 | ||||
| AK144155.1 | BAE25735.1 | |||
| BC002033.1 | 1 | 1285 | AAH02033.1 | |
| BC058180.1 | AAH58180.1 | |||
| U66887.1 | AAC52894.1 |
More Information
Set Membership: |
|

Genomic Map