Assay Details
Target Gene Details
Entrez Gene ID: | 279766 |
Gene Name: | rhomboid domain containing 3 |
Gene Aliases: |
Ptag |
Location: |
Chr.11:5095631-5106100 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rhbdd3 | NM_001290491.1 | 1 | 228 | NP_001277420.1 |
| NM_001290492.1 | 1 | 228 | NP_001277421.1 | |
| NM_177370.4 | 1 | 228 | NP_796344.1 | |
| AK053223.1 | 1 | 63 | ||
| AK077457.1 | 1 | 64 | BAC36809.1 | |
| AK156451.1 | 1 | 70 | BAE33717.1 | |
| BB841991.1 |
More Information
Set Membership: |
|

Genomic Map