Assay Details
Target Gene Details
Entrez Gene ID: | 114714 |
Gene Name: | RAD51 homolog C |
Gene Aliases: |
R51H3, Rad51l2 |
Location: |
Chr.11:87377607-87404954 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rad51c | NM_001291440.1 | NP_001278369.1 | ||
| NM_053269.3 | NP_444499.1 | |||
| AK076551.1 | BAC36388.1 | |||
| AK136204.1 | ||||
| BC141034.1 |
More Information
Set Membership: |
|

Genomic Map