Assay Details
Target Gene Details
Entrez Gene ID: | 279766 |
Gene Name: | rhomboid domain containing 3 |
Gene Aliases: |
Ptag |
Location: |
Chr.11:5095631-5106100 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rhbdd3 | NM_001290491.1 | NP_001277420.1 | ||
| NM_001290492.1 | NP_001277421.1 | |||
| NM_001290493.1 | NP_001277422.1 | |||
| NM_177370.4 | NP_796344.1 | |||
| NR_110961.1 | ||||
| AK053223.1 | ||||
| AK077457.1 | BAC36809.1 | |||
| AK156451.1 | BAE33717.1 | |||
| BC096693.1 | AAH96693.1 | |||
| BC132459.1 | ||||
| BC138479.1 | ||||
| BC145287.1 | ||||
| BC145289.1 |
More Information
Set Membership: |
|

Genomic Map