Assay Details
Target Gene Details
Entrez Gene ID: | 217344 |
Gene Name: | rhomboid 5 homolog 2 (Drosophila) |
Gene Aliases: |
4732465I17Rik, Rhbdl6, cub |
Location: |
Chr.11:116598165-116627019 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rhbdf2 | NM_001167680.1 | 12 | 1696 | NP_001161152.1 |
| NM_172572.3 | 12 | 1910 | NP_766160.2 | |
| AK028872.1 | 12 | 1911 | BAC26163.1 | |
| AK154639.1 | 12 | 1676 | ||
| AK171085.1 | 12 | 1692 | BAE42238.1 | |
| AY167043.1 | 12 | 1696 | AAO34122.1 | |
| BC031398.1 | 6 | 729 | AAH31398.1 | |
| BC052182.1 | 12 | 1644 | AAH52182.1 |
More Information
Set Membership: |
|

Genomic Map