Assay Details
Target Gene Details
Entrez Gene ID: | 77371 |
Gene Name: | Sec24 related gene family, member A (S. cerevisiae) |
Gene Aliases: |
- |
Location: |
Chr.11:51692263-51763627 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sec24a | NM_001290785.1 | NP_001277714.1 | ||
| NM_175255.3 | NP_780464.2 | |||
| AK029139.1 | BAC26319.1 | |||
| AK155182.1 | BAE33099.1 | |||
| BC117905.1 | ||||
| BC117906.1 |
More Information
Set Membership: |
|

Genomic Map