Assay Details
Target Gene Details
Entrez Gene ID: | 27029 |
Gene Name: | N-sulfoglucosamine sulfohydrolase (sulfamidase) |
Gene Aliases: |
4632406A19Rik |
Location: |
Chr.11:119343489-119355510 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sgsh | NM_018822.3 | 2 | 159 | NP_061292.2 |
| AF153827.1 | 2 | 133 | AAF29460.1 | |
| AF304053.1 | 2 | 133 | AAG41945.1 | |
| AF304054.1 | 2 | 133 | AAG41946.1 | |
| AK034220.1 | ||||
| AK052266.1 | 2 | 152 | BAC34906.1 | |
| AK076267.1 | 2 | 188 | ||
| AK128926.1 | 2 | 159 | BAC87657.1 | |
| AK141266.1 | 2 | 107 | BAE24624.1 | |
| BC130268.1 | 2 | 156 |
More Information
Set Membership: |
|

Genomic Map