Assay Details
Target Gene Details
Entrez Gene ID: | 27029 |
Gene Name: | N-sulfoglucosamine sulfohydrolase (sulfamidase) |
Gene Aliases: |
4632406A19Rik |
Location: |
Chr.11:119343489-119355510 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sgsh | NM_018822.3 | NP_061292.2 | ||
| AK034220.1 | ||||
| AK052266.1 | BAC34906.1 | |||
| AK076267.1 | ||||
| AK128926.1 | BAC87657.1 | |||
| BC130268.1 |
Target Gene Details
Entrez Gene ID: | 268512 |
Gene Name: | solute carrier family 26, member 11 |
Gene Aliases: |
F630021I08Rik |
Location: |
Chr.11:119355532-119381076 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc26a11 | NM_178743.3 | NP_848858.2 | ||
| AF345196.1 | AAO49173.1 | |||
| AK078545.1 | ||||
| AK089182.1 | BAC40782.1 | |||
| BC038604.1 | AAH38604.1 |
More Information
Set Membership: |
|

Genomic Map