Assay Details
Target Gene Details
Entrez Gene ID: | 268512 |
Gene Name: | solute carrier family 26, member 11 |
Gene Aliases: |
F630021I08Rik |
Location: |
Chr.11:119355532-119381076 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc26a11 | NM_178743.3 | NP_848858.2 | ||
| AF345196.1 | AAO49173.1 | |||
| AK078545.1 | ||||
| AK089182.1 | BAC40782.1 | |||
| BC038604.1 | AAH38604.1 |
More Information
Set Membership: |
|

Genomic Map