Assay Details
Target Gene Details
Entrez Gene ID: | 51799 |
Gene Name: | RUN domain containing 3A |
Gene Aliases: |
RPIP-8, Rap2ip, Rpip8 |
Location: |
Chr.11:102393403-102402939 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rundc3a | NM_001252347.1 | NP_001239276.1 | ||
| NM_016759.4 | NP_058039.1 | |||
| BC046319.1 | AAH46319.1 | |||
| BY258909.1 | ||||
| BY277835.1 | ||||
| U73941.1 | AAB51123.1 |

Genomic Map