Assay Details
Target Gene Details
Entrez Gene ID: | 22658 |
Gene Name: | polycomb group ring finger 2 |
Gene Aliases: |
Mel18, Rnf110, Zfp144, mel-18 |
Location: |
Chr.11:97688821-97701344 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcgf2 | NM_001163307.1 | NP_001156779.1 | ||
| NM_001163308.1 | NP_001156780.1 | |||
| NM_009545.2 | NP_033571.1 | |||
| AF483502.1 | AAL90776.1 | |||
| AF483503.1 | AAL90777.1 | |||
| AK142837.1 | ||||
| BC016419.1 | AAH16419.1 | |||
| BI991588.1 | ||||
| D90085.1 | BAA14122.1 |

Genomic Map