Assay Details
Target Gene Details
Entrez Gene ID: | 59040 |
Gene Name: | ras homolog gene family, member T1 |
Gene Aliases: |
2210403N23Rik, AA415293, AF244542, AI834919, Arht1, C430039G08Rik, Miro1 |
Location: |
Chr.11:80209005-80267907 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rhot1 | NM_001163354.1 | 13 | 1253 | NP_001156826.1 |
| NM_001163355.1 | 13 | 1253 | NP_001156827.1 | |
| NM_021536.7 | 13 | 1253 | NP_067511.4 | |
| AK019059.1 | 13 | 1213 | BAB31529.2 | |
| AK028597.1 | 13 | 1196 | BAC26025.1 | |
| AK031139.1 | 11 | 915 | BAC27274.1 | |
| AK032096.1 | 13 | 1226 | ||
| AK041119.1 | 13 | 1189 | BAC30828.1 | |
| AK043323.1 | ||||
| AK054472.1 | 13 | 1224 | BAC35792.1 | |
| AK078955.1 | 13 | 1196 | BAC37478.1 | |
| AK090071.1 | 6 | 579 | ||
| AK138830.1 | 13 | 1194 | BAE23792.1 | |
| AK141621.1 | 13 | 1080 | BAE24771.1 | |
| AK147749.1 | 13 | 1232 | BAE28113.1 | |
| AK147801.1 | 13 | 1258 | BAE28148.1 | |
| AK167502.1 | 13 | 1253 | BAE39578.1 | |
| AK169811.1 | 12 | 1910 | ||
| BC006918.1 | 4 | 344 | AAH06918.1 | |
| BC046785.1 | 13 | 1236 | AAH46785.1 | |
| BC052882.1 | 3 | 197 | AAH52882.1 | |
| BC058350.1 | 13 | 1168 | AAH58350.1 |
More Information
Set Membership: |
|

Genomic Map