Assay Details
Target Gene Details
Entrez Gene ID: | 20335 |
Gene Name: | SEC61, gamma subunit |
Gene Aliases: |
- |
Location: |
Chr.11:16501638-16508484 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sec61g | NM_001109971.1 | NP_001103441.1 | ||
| NM_001109972.1 | NP_001103442.1 | |||
| NM_011343.3 | NP_035473.2 | |||
| AK012393.1 | BAB28210.1 | |||
| AK012644.1 | BAB28376.1 | |||
| AK160390.1 | BAE35762.1 | |||
| AV456975.1 | ||||
| BC019158.1 | AAH19158.1 | |||
| BC081456.1 | AAH81456.1 | |||
| BY468217.1 | ||||
| U11027.1 | AAA19431.1 |
More Information
Set Membership: |
|

Genomic Map