Assay Details
Target Gene Details
Entrez Gene ID: | 11622 |
Gene Name: | aryl-hydrocarbon receptor |
Gene Aliases: |
Ah, Ahh, Ahre, In, bHLHe76 |
Location: |
Chr.12:35497979-35534989 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ahr | NM_013464.4 | 4 | 771 | NP_038492.1 |
| AF405560.1 | 4 | 405 | AAL89725.1 | |
| AF405561.1 | 4 | 405 | AAL89726.1 | |
| AF405562.1 | 4 | 405 | AAL89727.1 | |
| AF405563.1 | 4 | 405 | AAL89728.1 | |
| AF405565.1 | 4 | 405 | AAL89730.1 | |
| AF405566.1 | 4 | 405 | AAL89731.1 | |
| AF405567.1 | 4 | 405 | AAL89732.1 | |
| AF405568.1 | 4 | 405 | AAL89733.1 | |
| AF405570.1 | 4 | 405 | AAL89735.1 | |
| AK153678.1 | 4 | 772 | BAE32140.1 | |
| AK155380.1 | 4 | 823 | BAE33231.1 | |
| AY662080.1 | 4 | 405 | AAT75227.1 | |
| AY662081.1 | 3 | 301 | AAT75228.1 | |
| AY662082.1 | 4 | 405 | AAT75229.1 | |
| AY662083.1 | 4 | 405 | AAT75230.1 | |
| AY662084.1 | 4 | 405 | AAT75231.1 | |
| D38416.1 | 4 | 420 |
More Information
Set Membership: |
|

Genomic Map