Assay Details
Target Gene Details
Entrez Gene ID: | 217737 |
Gene Name: | AHA1, activator of heat shock protein ATPase 1 |
Gene Aliases: |
BC023857, p38 |
Location: |
Chr.12:87266484-87273998 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ahsa1 | NM_146036.1 | 2 | 295 | NP_666148.1 |
| AK076069.1 | 2 | 305 | BAC36160.1 | |
| AK077903.1 | 1 | 1536 | ||
| AK165289.1 | 2 | 315 | ||
| AK166644.1 | 2 | 268 | BAE38913.1 | |
| AK201587.1 | 2 | 109 | ||
| BC023857.1 | 2 | 262 | AAH23857.1 | |
| BC025552.1 | 2 | 295 | AAH25552.1 |
More Information
Set Membership: |
|

Genomic Map