Assay Details
Target Gene Details
Entrez Gene ID: | 217737 |
Gene Name: | AHA1, activator of heat shock protein ATPase 1 |
Gene Aliases: |
BC023857, p38 |
Location: |
Chr.12:87266484-87273998 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ahsa1 | NM_146036.1 | NP_666148.1 | ||
| AK076069.1 | BAC36160.1 | |||
| AK165289.1 | ||||
| AK166644.1 | BAE38913.1 | |||
| AK210244.1 | ||||
| BC023857.1 | AAH23857.1 | |||
| BC025552.1 | AAH25552.1 |
More Information
Set Membership: |
|

Genomic Map