Assay Details
Target Gene Details
Entrez Gene ID: | 238247 |
Gene Name: | AT rich interactive domain 4A (RBP1-like) |
Gene Aliases: |
A630009N03, A630067N03Rik, MmRBBP1, Rbbp1 |
Location: |
Chr.12:71015967-71099351 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arid4a | NM_001081195.1 | 17 | 1970 | NP_001074664.1 |
| AK149171.1 | 7 | 1030 | ||
| BC157975.1 | 16 | 1769 | ||
| BC158109.1 | 16 | 1772 |
More Information
Set Membership: |
|

Genomic Map