Assay Details
Target Gene Details
Entrez Gene ID: | 238247 |
Gene Name: | AT rich interactive domain 4A (RBP1-like) |
Gene Aliases: |
A630009N03, A630067N03Rik, MmRBBP1, Rbbp1 |
Location: |
Chr.12:71015967-71099351 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arid4a | NM_001081195.1 | NP_001074664.1 | ||
| AK042186.1 | 1 | 329 | BAC31192.1 | |
| AK050594.1 | BAC34332.1 | |||
| AK087354.1 | ||||
| AY116674.1 | AAN47196.1 | |||
| BC025436.1 | ||||
| BC058209.1 | AAH58209.1 |
More Information
Set Membership: |
|

Genomic Map