Assay Details
Target Gene Details
Entrez Gene ID: | 77577 |
Gene Name: | spinster homolog 3 |
Gene Aliases: |
9830002I17Rik |
Location: |
Chr.11:72498153-72550508 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spns3 | NM_029932.3 | 8 | 1118 | NP_084208.1 |
| AK020683.1 | 8 | 1119 | BAB32173.1 | |
| AK041376.1 | 7 | 1108 | BAC30922.1 | |
| AK156479.1 | 8 | 1382 | BAE33726.1 | |
| AK156781.1 | 8 | 1110 | ||
| BC150778.1 | 8 | 1101 |
More Information
Set Membership: |
|

Genomic Map