Assay Details
Target Gene Details
Entrez Gene ID: | 77097 |
Gene Name: | tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 2 |
Gene Aliases: |
3526402J09Rik, 5730590C14Rik |
Location: |
Chr.11:105589986-105929304 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tanc2 | NM_181071.3 | 20 | 3595 | NP_851416.2 |
| AK028312.1 | 4 | 441 | ||
| AK084601.1 | 3 | 370 | BAC39224.1 | |
| AK173111.1 | 11 | 2065 | BAD32389.1 | |
| BC089352.1 | 9 | 1586 | AAH89352.1 |

Genomic Map