Assay Details
Target Gene Details
Entrez Gene ID: | 77097 |
Gene Name: | tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 2 |
Gene Aliases: |
3526402J09Rik, 5730590C14Rik |
Location: |
Chr.11:105589986-105929304 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tanc2 | NM_181071.3 | NP_851416.2 | ||
| AK028312.1 | ||||
| AK084601.1 | 5 | 2145 | BAC39224.1 | |
| AK173111.1 | BAD32389.1 | |||
| BC089352.1 | AAH89352.1 |

Genomic Map