Assay Details
Target Gene Details
Entrez Gene ID: | 217356 |
Gene Name: | transmembrane channel-like gene family 8 |
Gene Aliases: |
EVIN2, Ever2, mFLJ00400 |
Location: |
Chr.11:117781598-117793137 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmc8 | NM_001195088.1 | 10 | 1471 | NP_001182017.1 |
| NM_001195089.1 | 10 | 1370 | NP_001182018.1 | |
| NM_001195090.1 | 10 | 1367 | NP_001182019.1 | |
| NM_181856.2 | 10 | 1367 | NP_862904.1 | |
| AK143862.1 | 10 | 1475 | ||
| AK156764.1 | 10 | 1328 | ||
| AK171772.1 | 10 | 1353 | BAE42658.1 | |
| AK220211.1 | 7 | 839 | BAD90136.1 | |
| AY236501.1 | 10 | 1279 | AAP69879.1 | |
| AY263160.1 | 9 | 1255 | AAP78775.1 | |
| BC138254.1 | 9 | 1243 | ||
| BC145210.1 | 9 | 1246 |

Genomic Map