Assay Details
Target Gene Details
Entrez Gene ID: | 193034 |
Gene Name: | transient receptor potential cation channel, subfamily V, member 1 |
Gene Aliases: |
OTRPC1, TRPV1alpha, TRPV1beta, VR-1, Vr1 |
Location: |
Chr.11:73231024-73261322 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trpv1 | NM_001001445.2 | 4 | 780 | NP_001001445.1 |
| AB180097.1 | 3 | 528 | BAD20301.1 | |
| AJ620495.1 | 3 | 568 | CAF05661.1 | |
| AK051315.1 | 4 | 638 | BAE20689.1 | |
| AY445519.1 | 3 | 528 | AAS01605.1 | |
| AY452083.1 | 3 | 561 | AAS15574.1 | |
| AY452084.1 | 3 | 561 | AAS15575.1 |
More Information
Set Membership: |
|

Genomic Map