Assay Details
Target Gene Details
Entrez Gene ID: | 22368 |
Gene Name: | transient receptor potential cation channel, subfamily V, member 2 |
Gene Aliases: |
GRC, OTRPC2, VRL-1, Vrl1 |
Location: |
Chr.11:62574486-62600305 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trpv2 | NM_011706.2 | 14 | 2410 | NP_035836.2 |
| AB021665.1 | 14 | 2492 | BAA78478.1 | |
| AK008689.1 | 12 | 1935 | ||
| AK089004.1 | 15 | 2425 | BAC40695.1 | |
| AK162841.1 | ||||
| AK180164.1 | 2 | 152 | ||
| AK209480.1 | 2 | 152 | ||
| AK212478.1 | 2 | 152 | ||
| BC005415.1 | 13 | 2357 | AAH05415.1 | |
| BY648241.1 | 1 | 66 |
More Information
Set Membership: |
|

Genomic Map