Assay Details
Target Gene Details
Entrez Gene ID: | 22368 |
Gene Name: | transient receptor potential cation channel, subfamily V, member 2 |
Gene Aliases: |
GRC, OTRPC2, VRL-1, Vrl1 |
Location: |
Chr.11:62574486-62600305 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trpv2 | NM_011706.2 | 16 | 2649 | NP_035836.2 |
| AB021665.1 | 16 | 2731 | BAA78478.1 | |
| AK008689.1 | 14 | 2174 | ||
| AK089004.1 | 17 | 2664 | BAC40695.1 | |
| AK162841.1 | ||||
| AK180164.1 | ||||
| BC005415.1 | 15 | 2596 | AAH05415.1 | |
| BY648241.1 | 3 | 305 |
More Information
Set Membership: |
|

Genomic Map