Assay Details
Target Gene Details
Entrez Gene ID: | 66132 |
Gene Name: | RIKEN cDNA 1110008L16 gene |
Gene Aliases: |
Kiaa0391, Mrpp3, mKIAA0391 |
Location: |
Chr.12:55302637-55382491 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1110008L16Rik | NM_025373.1 | NP_079649.1 | ||
| AK003589.1 | BAB22879.1 | |||
| AK031144.1 | BAC27278.1 | |||
| AK045434.1 | ||||
| AK151976.1 | BAE30844.1 | |||
| AK152028.1 | BAE30887.1 | |||
| AK172942.1 | BAD32220.1 | |||
| BC034876.1 | AAH34876.1 |

Genomic Map