Assay Details
Target Gene Details
Entrez Gene ID: | 66132 |
Gene Name: | RIKEN cDNA 1110008L16 gene |
Gene Aliases: |
Kiaa0391, Mrpp3, mKIAA0391 |
Location: |
Chr.12:55302637-55382491 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1110008L16Rik | NM_025373.1 | 6 | 2791 | NP_079649.1 |
| AK003589.1 | 6 | 721 | BAB22879.1 | |
| AK008749.1 | 2 | 217 | BAB25874.1 | |
| AK031144.1 | 6 | 2169 | BAC27278.1 | |
| AK045434.1 | 6 | 2159 | ||
| AK151976.1 | 6 | 2791 | BAE30844.1 | |
| AK152028.1 | 6 | 2791 | BAE30887.1 | |
| AK172942.1 | 6 | 595 | BAD32220.1 | |
| BC034876.1 | 6 | 1643 | AAH34876.1 |

Genomic Map