Assay Details
Target Gene Details
Entrez Gene ID: | 238247 |
Gene Name: | AT rich interactive domain 4A (RBP1-like) |
Gene Aliases: |
A630009N03, A630067N03Rik, MmRBBP1, Rbbp1 |
Location: |
Chr.12:71015967-71099351 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arid4a | NM_001081195.1 | 22 | 3722 | NP_001074664.1 |
| AK041444.1 | 1 | 113 | BAC30948.1 | |
| BC157975.1 | 21 | 3521 | ||
| BC158109.1 | 21 | 3524 |
More Information
Set Membership: |
|

Genomic Map