Assay Details
Target Gene Details
Entrez Gene ID: | 216987 |
Gene Name: | UTP6, small subunit (SSU) processome component, homolog (yeast) |
Gene Aliases: |
4732497O03Rik, HCA66 |
Location: |
Chr.11:79932320-79962426 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Utp6 | NM_144826.3 | 8 | 663 | NP_659075.1 |
| AF334387.1 | 8 | 990 | AAO15383.1 | |
| AK029151.1 | 8 | 664 | BAC26328.1 | |
| AK084976.1 | 8 | 656 | BAC39327.1 | |
| AK164010.1 | 8 | 668 | ||
| AK169785.1 | 8 | 670 | BAE41364.1 | |
| BC018250.1 | 8 | 627 | AAH18250.1 | |
| BC023920.1 | 8 | 657 | AAH23920.1 | |
| BC024850.1 | 8 | 658 | AAH24850.1 |
More Information
Set Membership: |
|

Genomic Map