Assay Details
Target Gene Details
Entrez Gene ID: | 103724 |
Gene Name: | TBC1 domain family, member 10a |
Gene Aliases: |
AI447804, EPI64, Tbc1d10, mFLJ00288 |
Location: |
Chr.11:4186833-4215507 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d10a | NM_134023.1 | NP_598784.1 | ||
| AK140872.1 | BAE24505.1 | |||
| AK208062.1 | ||||
| BC018300.1 | AAH18300.1 | |||
| BC029773.1 | AAH29773.1 | |||
| BC037230.1 | AAH37230.1 |
More Information
Set Membership: |
|

Genomic Map