Assay Details
Target Gene Details
Entrez Gene ID: | 207592 |
Gene Name: | TBC1 domain family, member 16 |
Gene Aliases: |
B930087K01, BC026530, Tcd1d16 |
Location: |
Chr.11:119143043-119228499 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d16 | NM_172443.3 | 4 | 978 | NP_766031.2 |
| AK047555.1 | 4 | 979 | BAC33084.1 | |
| BC057634.1 | 4 | 940 | AAH57634.1 | |
| BQ831250.1 |
More Information
Set Membership: |
|

Genomic Map