Assay Details
Target Gene Details
Entrez Gene ID: | 338369 |
Gene Name: | transmembrane protein 220 |
Gene Aliases: |
- |
Location: |
Chr.11:67025154-67035312 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem220 | NM_001291042.1 | NP_001277971.1 | ||
| NM_001291043.1 | NP_001277972.1 | |||
| NM_177392.3 | NP_796366.1 | |||
| AK043094.1 | BAC31457.1 | |||
| AK078505.1 | BAC37312.1 | |||
| BC110628.1 | AAI10629.1 | |||
| BC110629.1 | AAI10630.1 |
More Information
Set Membership: |
|

Genomic Map