Assay Details
Target Gene Details
Entrez Gene ID: | 11431 |
Gene Name: | acid phosphatase 1, soluble |
Gene Aliases: |
4632432E04Rik, AI427468, Acp-1, LMW-PTP |
Location: |
Chr.12:30893326-30911621 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acp1 | NM_001110239.1 | NP_001103709.1 | ||
| NM_021330.4 | NP_067305.2 | |||
| AK014603.1 | BAB29458.1 | |||
| AK018329.1 | BAB31164.1 | |||
| AK019186.1 | BAB31593.1 | |||
| AK082955.1 | BAC38707.1 | |||
| AK136144.1 | BAE22844.1 | |||
| AK152429.1 | BAE31211.1 | |||
| AK167887.1 | BAE39900.1 | |||
| AK170284.1 | BAE41686.1 | |||
| BC039744.1 | AAH39744.1 | |||
| BC093527.1 | AAH93527.1 | |||
| BC096368.1 | AAH96368.1 | |||
| BC111893.1 | ||||
| BC112413.1 | ||||
| BC132258.1 | ||||
| BC145199.1 | ||||
| Y17343.1 | CAA76753.1 | |||
| Y17345.1 | CAA76755.1 |
More Information
Set Membership: |
|

Genomic Map