Assay Details
Target Gene Details
Entrez Gene ID: | 76795 |
Gene Name: | TBC1 domain family, member 9B |
Gene Aliases: |
- |
Location: |
Chr.11:50131057-50172785 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d9b | NM_001290759.1 | 16 | 2757 | NP_001277688.1 |
| NM_001290760.1 | 16 | 2760 | NP_001277689.1 | |
| NM_029745.2 | 16 | 2760 | NP_084021.2 | |
| AK031992.1 | 1 | 179 | BAC27640.1 | |
| AK035457.1 | 16 | 2725 | BAC29068.1 | |
| AK049999.1 | 16 | 2720 | BAC34024.2 | |
| AK088860.1 | 11 | 1710 | ||
| AK147964.1 | 16 | 2725 | BAE28254.1 | |
| AK153522.1 | 1 | 2030 | ||
| AK172999.1 | 17 | 2697 | BAD32277.1 | |
| AK180139.1 | ||||
| AK181925.1 | ||||
| AK183545.1 | ||||
| AK187126.1 | ||||
| AK188982.1 | ||||
| AK192419.1 | ||||
| AK192550.1 | ||||
| AK195739.1 | ||||
| AK196195.1 | ||||
| AK197239.1 | ||||
| AK197459.1 | ||||
| AK215906.1 | 1 | 107 | ||
| BC048085.1 | 7 | 1074 | AAH48085.1 | |
| BC058596.1 | 3 | 294 | AAH58596.1 | |
| BC062928.1 | 16 | 2747 | AAH62928.1 | |
| BC065080.1 | 16 | 2739 | AAH65080.1 |
More Information
Set Membership: |
|

Genomic Map