Assay Details
Target Gene Details
Entrez Gene ID: | 76795 |
Gene Name: | TBC1 domain family, member 9B |
Gene Aliases: |
- |
Location: |
Chr.11:50131057-50172785 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d9b | NM_001290759.1 | NP_001277688.1 | ||
| NM_001290760.1 | NP_001277689.1 | |||
| NM_029745.2 | NP_084021.2 | |||
| AK031992.1 | BAC27640.1 | |||
| AK035457.1 | BAC29068.1 | |||
| AK049999.1 | BAC34024.2 | |||
| AK088860.1 | ||||
| AK147964.1 | BAE28254.1 | |||
| AK172999.1 | BAD32277.1 | |||
| AK184661.1 | ||||
| AK186352.1 | ||||
| AK188575.1 | ||||
| AK188975.1 | ||||
| AK192140.1 | ||||
| AK193080.1 | ||||
| AK194602.1 | ||||
| AK195900.1 | ||||
| AK199606.1 | ||||
| AK207865.1 | ||||
| AK207874.1 | ||||
| AK207903.1 | ||||
| AK210062.1 | ||||
| AK210511.1 | ||||
| AK210947.1 | ||||
| AK212277.1 | ||||
| AK214457.1 | ||||
| AK214532.1 | ||||
| AK218626.1 | ||||
| AK219161.1 | ||||
| BC048085.1 | AAH48085.1 | |||
| BC058596.1 | AAH58596.1 | |||
| BC062928.1 | AAH62928.1 | |||
| BC065080.1 | AAH65080.1 |
More Information
Set Membership: |
|

Genomic Map